Acetic acid 1.00 N

Supplier: Ricca Chemical

Synonyms: Methanecarboxylic acidVinegar acidEthanoic acid

R0150000-4A R0150000-1A R0150000-20F
RC150-1EA 156.58 USD
RC150-1 RC150-32 RC150-5
Acetic acid 1.00 N
Acetic acid
Formula: H₃CCOOH
MW: 60.05 g/mol
MDL Number: MFCD00036152
CAS Number: 64-19-7
UN: 2789
ADR: 8,II
https://us.vwr.com/stibo/web/std.lang.all/31/15/723115.jpg

Order Now

Specification Test Results

Appearance Clear, colorless, mild vinegar odor
Assay at 20 °C (traceable to NIST SRM 84) 1.000 ± 0.002 N at 20.0 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR