Bromothymol blue-phenol red mixed indicator
Supplier: Ricca Chemical
R1385000-500A
R1385000-120A
RC1385-16EA
74
USD
RC1385-16
RC1385-4
Bromothymol blue-phenol red mixed indicator
Bromothymol blue-phenol red
Specification Test Results
Appearance | Dark green to purplish red, odorless |
pH at 25 °C (traceable to NIST SRM 186 & 191) | 7.50 ± 0.05 pH at 25.0 °C |
Suitability for Use | Yellow (pH 7.5) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...