Sodium chloride 99.0-100.5% USP, Ph. Eur., JP, BP, FCC (GMP)
Supplier: Macco Organiques S.R.O
Synonyms:
Halite
, Sea salt
, Saline
For excipient use only.
Sodium chloride is in conformity with the last edition of:
European Pharmacopoeia,
American Pharmacopoeia,
Japanese Pharmacopoeia,
British pharmacopoeia,
Food Chemical Codex
No material of animal or human origin is used in the raw materials, in the production and distribution process of this product.
This product is BSE/TSE, GMO, allergen free.
Certifications: GMP for active substances, ISO 9001:2000, CEP, Kosher.
Ordering information: Minimum order quantities may apply; contact your VWR representative for more information.
Formula:
NaCl MW: 58.44 g/mol Boiling Pt: 1413 °C (1013 hPa) Melting Pt: 801 °C Density: 2.16 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00003477 CAS Number: 7647-14-5 EINECS: 231-598-3 Merck Index: 14,08599 |
Specification Test Results
Identity A) Sodium (PhEur/BP/USP/FCC/JP) | Pass test |
Identity B) Chlorides (PhEur/BP/USP/FCC/JP) | Pass test |
Assay (PhEur/BP/USP/FCC/JP) | 99.0-100.5 |
Appearance (PhEur/BP) | White or almost white, crystalline powder or colourless crystals or white or almost white perls. |
Solubility (PhEur) | Freely soluble in water, practically insoluble in anhydrous ethanol. |
Appearance of solution (PhEur/BP/USP/JP) | Clear and colorless |
Acidity or alkalinity (PhEur/BP/USP/JP) | Corresponds to test |
Bromides (PhEur/BP/USP) | 0.01% Max. |
Ferrocyanides (PhEur/BP/USP/JP) | Corresponds to test |
Iodides (PhEur/BP/USP/JP) | Corresponds to test |
Nitrites (PhEur/BP/USP) | Corresponds to test |
Phosphates (PhEur/BP/USP) | 0.0025 % Max. |
Sulphates (PhEur/BP/USP) | 0.0025 % Max. |
Aluminium (PhEur/BP/USP) | 0.00002 % Max. |
Arsenic (PhEur/BP/USP) | 0.0001 % Max. |
Barium (PhEur/BP/USP/JP) | Corresponds to test |
Iron (PhEur/BP/USP) | 0.0002 % Max. |
Magnesium and alkaline- earth metals (PhEur/BP/USP) | 0.01 % (calculated as Ca) Max. |
Potassium (PhEur/BP/USP) | 0.05% Max. |
Heavy metals (PhEur/BP/USP) | 0.0005% Max. |
Loss on drying (PhEur/BP/USP/FCC/JP) | 0.5% Max. |
Bacterial endotoxines (PhEur/BP) | 5 I.U. per gram |
Residual solvents (USP) | corresponds to test |
Appearance (FCC) | White crystalline solid of variable particle size. |
Iron (FCC) | 0.0016% Max. |
Sodium Ferrocyanide (FCC) | 0.0013% Max |
Iodine (FCC) | 0.006 - 0.010 % KI |
Calcium and magnesium (FCC) | 0.9 % Salt other than evaporated salt max. 0.35 % Evaporated salt Max. |
Appearance (JP) | Colorless or white crystals or crystalline powder. |
Solubility (JP) | Freely soluble in water, practically insoluble in ethanol (99.5). |
Bromides (JP) | Corresponds to test |
Phosphates (JP) | Corresponds to test |
Arsenic (JP) | 0.0002% Max. |
Iron (JP) | Corresponds to test |
Magnesium and alkaline-earth metals (JP) | Corresponds to test |
Heavy metals (JP) | 0.0003% Max. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...