Sodium bisulfite, dried, purified, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
|
Danger
|
Synonyms:
Sodium hydrogen sulfite
, Sodium hydrogen sulphite
Management of Change (MOC) category = N
BAKR3557-01
BAKR3557-01
JT3557-1CS
938.13
USD
CAJT3557-1
JT3557-1
Sodium bisulfite, dried, purified, J.T.Baker®
Sodium hydrogen sulphite
Formula:
HNaO₃S MW: 104.06 g/mol Melting Pt: 300 °C Density: 2.476 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00167602 CAS Number: 7631-90-5 EINECS: 231-548-0 UN: 2693 ADR: 8,III Merck Index: 12,08731 |
Specification Test Results
For Laboratory,Research,or Manufacturing Use | |
Appearance | Passes Test |
Identification | Passes Test |
Solubility in H₂O | Passes Test |
Thiosulfate (S₂O₃) | Passes Test |
Heavy Metals (as Pb) | ≤0.003 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...