di-Potassium oxalate monohydrate 98.5-101.0%, GR ACS, Supelco®

Warning

Synonyms: Oxalic acid potassium salt monohydrate, Potassium oxalate hydrate, Potassium oxalate monohydrate, Potassium ethanedioate monohydrate

EM-PX1535-1CS 461.66 USD
EM-PX1535-1
di-Potassium oxalate monohydrate 98.5-101.0%, GR ACS, Supelco®
di-Potassium oxalate monohydrate
Formula: (COOK)₂·H₂O
MW: 184.23 g/mol
Melting Pt: 356
Density: 2.154 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00150033
CAS Number: 6487-48-5
EINECS: 209-506-8
UN: 2811
ADR: 6.1 III
Merck Index: 12,07820

Order Now

Specification Test Results

Assay 98.5-101.0%
Ammonium (NH4) 0.002% max
Chloride (Cl) 0.002% max
Color White
Form Crystals
Heavy metals (as Pb) 0.002% max
Insoluble matter 0.01% max
Iron (Fe) 0.001% max
Neutrality To pass test
Sodium (Na) 0.02% max
Substances darkened by hot sulfuric acid To pass test
Sulfate (SO4) 0.01% max


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR