Piperidine, Supelco®
Supplier: MilliporeSigma
|
Danger
|
Synonyms:
Hexahydropyridine
, Hexazane
, Pentamethyleneimine
PX1235-8
PX1235-4000
EM-PX1235-8EA
3195
USD
EM-PX1235-8
89423-634
Piperidine, Supelco®
Piperidine
Formula:
C₅H₁₁N MW: 85.15 g/mol Boiling Pt: 107.3 °C (1013 hPa) Melting Pt: –10 °C Density: 0.8622 g/cm³ (20 °C) Flash Pt: 16 °C Storage Temperature: Ambient |
MDL Number:
MFCD00005979 CAS Number: 110-89-4 EINECS: 203-813-0 UN: 2401 ADR: 8,I Merck Index: 12,07621 |
Specification Test Results
Assay (GC) | 99.5% min |
Color (APHA) | 20 max |
Form | Clear liquid |
Identity (IR-spectrum) | Conforms |
Water (H2O) | 0.2% max |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...