Plantcon™ has been designed to reduce the time spent preparing an in vitro growth environment for plant cultures and to minimize the risk of contamination.
- Aseptic germination of seeds or spores.
- Stage I production of clean explant for multiplication of callus formation.
- Stage II multiplication of plants through enhanced axillary branching, adventitious buds, embryoids or callus.
- Stage III pretransplant rooting and hardening of plantlets for transfer to soil.
- As a shipping container for pretransplant plantlets. PlantCon™ containers may be used with liquid or agar solidified media. They can be used for many plant cell culture procedures that currently utilize glass containers.
The PlantCon™ container consists of a cover and a bottom, and features a vent for gas exchange, and a drip control lip to recycle condensation
PlantCon™ is a line of presterilized, disposable, plastic growth containers. This labware has been designed to reduce the time spent preparing an in vitro growth environment for plant cultures and to minimize the risk of contamination. Instead the PlantCon™ system permits you to start culturing plant cells and plantlets with the following advantages: Unbreakable -- Safe to use, Presterilized -- Easy to use, Clear plastic -- Excellent light transmission, Disposable -- Eliminates messy glass cleanup, Space Saving -- Stackable, Manufactured under strict quality control procedures, Scientifically designed for plant cell culture, Special cover positioning -- Special lugs in the cover lip allow the user to snap on the cover tight for handling or loose for free exchange of atmospheres.
PlantCon™ containers may be used with liquid or agar solidified media. They can be used for many plant cell culture procedures that currently utilize glass containers.
The cover can be snapped on tight, for long-term culture, or loosely fitted during acclimation to external conditions prior to transplantation. The covers and bottoms are sterile and nested separately in a sleeve of 25 each. Provided presterilized, made of clear unbreakable plastic. These disposable, space saving containers are specifically designed for optimal plant cell culture applications.