Vernier® Projectile Launcher
Supplier: Vernier Software & Technology
VPL GGL-VPL
470134-258EA 439.99 USD
470134-258 470303-822
Vernier® Projectile Launcher
Educational Materials Physics Educational Materials Mechanics Learning Activities
Students use the Vernier Projectile Launcher to investigate important concepts in two-dimensional kinematics.
  • Students Investigate Important Concepts in Two-Dimensional Kinematics
  • Device Plugs into Compatible Data-Collection Interface
  • Pneumatic Launching System
  • Heavy Base
  • Built-In Photogates
They can launch steel balls at angles between 0 and 70° and over distances up to 2.5 m. The solid, heavy base provides an easy-to-use and reliable setup. A unique pneumatic launching system provides excellent repeatability and allows users to set the launch speed.
More Product Information