HyClone™ HyQ-RS Reduced Serum Media, HyClone products (Cytiva)

Supplier: Cytiva

SH30565.02 SH30564.01 SH30565.01
82017-968EA 46.62 USD
82017-968 82017-970 82017-966
HyClone™ HyQ-RS Reduced Serum Media, HyClone products (Cytiva)
Media for Microbiology Prepared Media for Microbiology

HyClone HyQ-RS Reduced Serum Media are nutritionally enhanced versions of classical media requiring a lower concentration of serum supplementation


  • Reduce FBS supplementation rate to 2 to 4% for most cell culture applications
  • Suitable for a wide variety of cell types normally used with standard classical formulations
  • Maintain cell performance while reducing serum supplementation by up to 80%
  • All contain phenol red
  • 0,1 μm sterile filtered
  • Manufactured using ISO 9001:2000 certified processes


We can determine which (if any) ingredients in a medium formulation are of animal origin or, if they have come in contact with animal derived materials in manufacture.


High yield cultures begin with high quality media. In order to maintain these standards, all HyClone products as well as rea material components must pass strict Quality Control testing to confirm the appropriate level of quality. Each raw material has a specification document describing the product, the required grade or purity, the source, and the criteria for acceptance on incoming inspection.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR