HyClone™ Fetal Bovine Serum (United States), Super Low IgG, HyClone Products (Cytiva)
Supplier: Cytiva
SH30898.02 SH30898.03 SH30898.03HI SH30898.03IR25-40 3089803IH25-40 SH30898.01 SH30898.02HI
95042-942EA 301.36 USD
95042-942 95042-944 95042-948 76324-912 76324-914 76534-640 76635-870
HyClone™ Fetal Bovine Serum (United States), Super Low IgG, HyClone Products (Cytiva)
Sera (Serum)
Enhance cell culture growth with U.S.-sourced Cytiva HyClone Super Low IgG Fetal Bovine Serum (FBS).
  • Processed via protein G chromatography
  • Enhance cell culture growth
  • U.S. sourced
Enhance cell culture growth with U.S.-sourced Cytiva HyClone Super Low IgG Fetal Bovine Serum (FBS), processed via protein G chromatography to substantially reduce immunoglobulin G (IgG) levels.