Bottom Drip Adapter for Big Columns, Two Filters, Ace Glass Incorporated
Supplier: Ace Glass
5857-86 5857-89
89053-038EA 722.05 USD
89053-038 89053-040
Bottom Drip Adapter for Big Columns, Two Filters, Ace Glass Incorporated
Adapters Adapters, Joints with Tube Connectors
This end-fitting adapter fits large columns with #80 Ace-Thred connections.
  • Recessed
  • Two polyethylene filter discs
  • Bottom outlet thread accepts 1/2-inch or 3/8-inch female NPT fitting
Adapter is recessed to allow the perforated support plate and retaining ring to fit. Bottom outlet is a thread that will accept a 1/2-inch or 3/8-inch female NPT fitting.
Ordering information: Complete item includes adapter, size -235 FETFE o-ring, 89052-914 perforated glass support plate, PTFE (89053-020) retaining ring, 100 and 350 micron polyethylene filter discs.