(5-Amino-2-hydroxymethylphenyl)boronic acid hydrochloride, dehydrated

Supplier: Matrix Scientific

Synonyms: 5-Amino-2-(hydroxymethyl)benzeneboronic acid dehydrate hydrochloride, 6-Aminobenzo[c][1,2]oxaborol-1(3H)-ol hydrochloride

069285-1G 069285-5G
101939-570EA 501.54 USD
101939-570 101939-572
(5-Amino-2-hydroxymethylphenyl)boronic acid hydrochloride, dehydrated
(5-Amino-2-hydroxymethylphenyl)boronic acid hydrochloride

Hazard: Irritant

Formula: C₇H₉BClNO₂
MW: 185.42 g/mol
MDL Number: MFCD04115645
CAS Number: 117098-93-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR