3-(Fluoromethyl)piperidine hydrochloride ≥95%

Supplier: Matrix Scientific

069005-500MG 069005-1G 069005-5G
101938-602EA 581.54 USD
101938-602 101938-604 101938-606
3-(Fluoromethyl)piperidine hydrochloride ≥95%
3-(Fluoromethyl)piperidine hydrochloride

Hazard: Irritant

Formula: C₆H₁₃ClFN
MDL Number: MFCD17015069
CAS Number: 1241725-60-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR