(S)-(+)-1-Boc-3-(aminomethyl)pyrrolidine ≥95%

Supplier: Matrix Scientific

Synonyms: (S)-(+)-1-Boc-3-(aminomethyl)pyrrolidine, (S)-1-Boc-3-(Aminomethyl)pyrrolidine, (S)-3-(Aminomethyl)-1-(tert-butoxycarbonyl)pyrrolidine, (S)-3-(Aminomethyl)-1-Boc-pyrrolidine, tert-Butyl (S)-3-(aminomethyl)-1-pyrrolidinecarboxylate

068628-1G 068628-5G
101936-986EA 144.62 USD
101936-986 101936-988
(S)-(+)-1-Boc-3-(aminomethyl)pyrrolidine ≥95%
(S)-(+)-1-Boc-3-(aminomethyl)pyrrolidine

Hazard: Irritant

Formula: C₁₀H₂₀N₂O₂
MW: 200.282 g/mol
MDL Number: MFCD02179397
CAS Number: 199175-10-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR