(R)-1-Methyl-3-pyrrolidinol ≥95%

Supplier: Matrix Scientific

Synonyms: (R)-3-Hydroxy-1-methylpyrrolidine, (R)-(-)-1-Methyl-3-pyrrolidinol, (R)-3-Hydroxy-1-methyl-pyrrolidine

068623-1G 068623-5G
101936-964EA 176.92 USD
101936-964 101936-966
(R)-1-Methyl-3-pyrrolidinol ≥95%
(R)-1-Methyl-3-pyrrolidinol

Hazard: Irritant

Formula: C₅H₁₁NO
MDL Number: MFCD03788747
CAS Number: 104641-60-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR