2-Amino-5-hydroxymethylpyridine ≥95%

Supplier: Matrix Scientific

Synonyms: 2-Amino-5-pyridinemethanol, 6-Amino-3-pyridinemethanol, 2-Amino-5-(hydroxymethyl)pyridine, 2-Aminopyridine-5-methanol, (6-Aminopyridin-3-yl)methanol, (6-Amino-3-pyridyl)methanol

068449-500MG 068449-1G 068449-5G
101936-194EA 172.31 USD
101936-194 101936-196 101936-198
2-Amino-5-hydroxymethylpyridine ≥95%
2-Amino-5-hydroxymethylpyridine

Hazard: Irritant

Formula: C₆H₈N₂O
MW: 124.14 g/mol
MDL Number: MFCD06659089
CAS Number: 113293-71-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR