4-Amino-2-bromobenzonitrile ≥98%

Supplier: Matrix Scientific

066623-5G 066623-25G
101931-998EA 29.23 USD
101931-998 101932-000
4-Amino-2-bromobenzonitrile ≥98%
4-Amino-2-bromobenzonitrile

TCA listed: T
Hazard: Toxic

Formula: C₇H₅BrN₂
MW: 197.03 g/mol
MDL Number: MFCD03695814
CAS Number: 53312-82-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR