4-Amino-6-chlorobenzene-1,3-disulfonamide

Supplier: Matrix Scientific

Synonyms: 4-Amino-6-chloro-1,3-benzenedisulphonamide, 4-Amino-6-chloro-1,3-benzenedisulfonamide

058352-5G 058352-25G
101914-638EA 15.38 USD
101914-638 101914-640
4-Amino-6-chlorobenzene-1,3-disulfonamide
4-Amino-6-chlorobenzene-1,3-disulphonamide

Hazard: Irritant

Formula: C₆H₈ClN₃O₄S₂
MW: 285.7300 g/mol
MDL Number: MFCD00007933
CAS Number: 121-30-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR