5-Fluoro-4-(trimethylsilyl)-1H-pyrrolo[2,3-b]pyridine

Supplier: Matrix Scientific

055638-100MG
101909-832EA 364.62 USD
101909-832
5-Fluoro-4-(trimethylsilyl)-1H-pyrrolo[2,3-b]pyridine
5-Fluoro-4-(trimethylsilyl)-1H-pyrrolo[2,3-b]pyridine

Hazard: Irritant

Formula: C₁₀H₁₃FN₂Si
MW: 208.31 g/mol
MDL Number: MFCD16628250
CAS Number: 1228665-73-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR