2-(3-((tert-Butyldimethylsilyloxy)methyl)pyrrolidin-1-yl)-6-(pyrrolidin-1-yl)pyridine

Supplier: Matrix Scientific

055613-100MG
101909-782EA 364.62 USD
101909-782
2-(3-((tert-Butyldimethylsilyloxy)methyl)pyrrolidin-1-yl)-6-(pyrrolidin-1-yl)pyridine
2-(3-((tert-Butyldimethylsilyloxy)methyl)pyrrolidin-1-yl)-6-(pyrrolidin-1-yl)pyridine

Hazard: Irritant

Formula: C₂₀H₃₅N₃OSi
MDL Number: MFCD15530311
CAS Number: 1228666-52-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR