2-Amino-5-fluoronicotinic acid

Supplier: Matrix Scientific

055559-100MG
101909-674EA 592.31 USD
101909-674
2-Amino-5-fluoronicotinic acid
2-Amino-5-fluoronicotinic acid

Hazard: Irritant

Formula: C₆H₅FN₂O₂
MDL Number: MFCD13190373
CAS Number: 1196154-03-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR