N'-Hydroxy-2-[(2-hydroxyethyl)(methyl)amino]benzenecarboximidamide

Supplier: Matrix Scientific

052612-500MG 052612-2.5G
101900-334EA 364.62 USD
101900-334 101900-336
N'-Hydroxy-2-[(2-hydroxyethyl)(methyl)amino]benzenecarboximidamide
N'-Hydroxy-2-[(2-hydroxyethyl)(methyl)amino]benzenecarboximidamide

Hazard: Irritant

Formula: C₁₀H₁₅N₃O₂
MDL Number: MFCD12772602
CAS Number: 1216330-09-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR