2-{4-[3-Amino-4-(methylsulfonyl)phenyl]-1-piperazinyl}-1-ethanol

Supplier: Matrix Scientific

052520-500MG 052520-2.5G
101899-966EA 364.62 USD
101899-966 101899-968
2-{4-[3-Amino-4-(methylsulfonyl)phenyl]-1-piperazinyl}-1-ethanol
2-{4-[3-Amino-4-(methylsulfonyl)phenyl]-1-piperazinyl}-1-ethanol

Hazard: Irritant

Formula: C₁₃H₂₁N₃O₃S
MDL Number: MFCD13562254
CAS Number: 1219957-14-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR