N1-Cyclohexyl-N1-methyl-4-(methylsulfonyl)-1,3-benzenediamine
Supplier: Matrix Scientific
052516-500MG
052516-2.5G
101899-950EA
364.62
USD
101899-950
101899-952
N1-Cyclohexyl-N1-methyl-4-(methylsulfonyl)-1,3-benzenediamine
N1-Cyclohexyl-N1-methyl-4-(methylsulfonyl)-1,3-benzenediamine
Hazard: Irritant
Formula:
C₁₄H₂₂N₂O₂S MW: 282.41 g/mol |
MDL Number:
MFCD13562250 CAS Number: 1220033-81-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...