1-[3-Amino-4-(methylsulfonyl)phenyl]-4-piperidinol

Supplier: Matrix Scientific

052514-500MG 052514-2.5G
101899-942EA 364.62 USD
101899-942 101899-944
1-[3-Amino-4-(methylsulfonyl)phenyl]-4-piperidinol
1-[3-Amino-4-(methylsulfonyl)phenyl]-4-piperidinol

Hazard: Irritant

Formula: C₁₂H₁₈N₂O₃S
MW: 270.35 g/mol
MDL Number: MFCD13562248
CAS Number: 1219957-10-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR