N-Benzyl-N-ethyl-4-piperidinecarboxamide hydrochloride

Supplier: Matrix Scientific

052501-500MG 052501-2.5G
101899-890EA 364.62 USD
101899-890 101899-892
N-Benzyl-N-ethyl-4-piperidinecarboxamide hydrochloride
N-Benzyl-N-ethyl-4-piperidinecarboxamide hydrochloride

Hazard: Irritant

Formula: C₁₅H₂₃ClN₂O
MW: 282.82 g/mol
MDL Number: MFCD13186579
CAS Number: 6308-67-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR