N,N-Dipropyl-4-piperidinecarboxamide hydrochloride

Supplier: Matrix Scientific

052492-500MG 052492-2.5G
101899-854EA 364.62 USD
101899-854 101899-856
N,N-Dipropyl-4-piperidinecarboxamide hydrochloride
N,N-Dipropyl-4-piperidinecarboxamide hydrochloride

Hazard: Irritant

Formula: C₁₂H₂₅ClN₂O
MW: 248.8 g/mol
MDL Number: MFCD13562229
CAS Number: 1220033-73-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR