N-Methyl-N-(tetrahydro-2H-pyran-4-ylmethyl)-3-piperidinecarboxamide hydrochloride

Supplier: Matrix Scientific

052475-500MG 052475-2.5G
101899-786EA 364.62 USD
101899-786 101899-788
N-Methyl-N-(tetrahydro-2H-pyran-4-ylmethyl)-3-piperidinecarboxamide hydrochloride
N-Methyl-N-(tetrahydro-2H-pyran-4-ylmethyl)-3-piperidinecarboxamide hydrochloride

Hazard: Irritant

Formula: C₁₃H₂₅ClN₂O₂
MDL Number: MFCD13562220
CAS Number: 1219972-70-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR