N,N-Diallyl-2-pyrrolidinecarboxamide hydrochloride

Supplier: Matrix Scientific

052398-500MG 052398-2.5G
101899-478EA 364.62 USD
101899-478 101899-480
N,N-Diallyl-2-pyrrolidinecarboxamide hydrochloride
N,N-Diallyl-2-pyrrolidinecarboxamide hydrochloride

Hazard: Irritant

Formula: C₁₁H₁₉ClN₂O
MDL Number: MFCD13562155
CAS Number: 1246172-86-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR