3-Amino-1-[3,4-dihydro-1(2H)-quinolinyl]-1-propanone hydrochloride

Supplier: Matrix Scientific

052299-500MG 052299-2.5G
101899-082EA 364.62 USD
101899-082 101899-084
3-Amino-1-[3,4-dihydro-1(2H)-quinolinyl]-1-propanone hydrochloride
3-Amino-1-[3,4-dihydro-1(2H)-quinolinyl]-1-propanone hydrochloride

Hazard: Irritant

Formula: C₁₂H₁₇ClN₂O
MDL Number: MFCD11857967
CAS Number: 1170377-26-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR