3-Amino-N,N-dipropylpropanamide hydrochloride
Supplier: Matrix Scientific
052291-500MG
052291-2.5G
101899-050EA
364.62
USD
101899-050
101899-052
3-Amino-N,N-dipropylpropanamide hydrochloride
3-Amino-N,N-dipropylpropanamide hydrochloride
Hazard: Irritant
Formula:
C₉H₂₁ClN₂O MW: 208.73 g/mol |
MDL Number:
MFCD13562052 CAS Number: 1220033-36-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...