5-Bromo-N-methyl-N-(tetrahydro-2H-pyran-4-ylmethyl)-2-pyridinamine
Supplier: Matrix Scientific
052159-500MG
052159-2.5G
101898-526EA
364.62
USD
101898-526
101898-528
5-Bromo-N-methyl-N-(tetrahydro-2H-pyran-4-ylmethyl)-2-pyridinamine
5-Bromo-N-methyl-N-(tetrahydro-2H-pyran-4-ylmethyl)-2-pyridinamine
Hazard: Irritant
Formula:
C₁₂H₁₇BrN₂O MW: 285.19 g/mol |
MDL Number:
MFCD13561948 CAS Number: 1220017-48-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...