5-Bromo-2-(piperidin-1-yl)pyridine
Supplier: Matrix Scientific
052135-500MG
052135-2.5G
101898-430EA
364.62
USD
101898-430
101898-432
5-Bromo-2-(piperidin-1-yl)pyridine
5-Bromo-2-(piperidin-1-yl)pyridine
Hazard: Irritant
Formula:
C₁₀H₁₃BrN₂ |
MDL Number:
MFCD00234654 CAS Number: 24255-95-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...