4-Chloro-6-(4-methyl-1-piperidinyl)-1,3,5-triazin-2-amine
Supplier: Matrix Scientific
052085-500MG
052085-2.5G
101898-230EA
364.62
USD
101898-230
101898-232
4-Chloro-6-(4-methyl-1-piperidinyl)-1,3,5-triazin-2-amine
4-Chloro-6-(4-methyl-1-piperidinyl)-1,3,5-triazin-2-amine
Hazard: Irritant
Formula:
C₉H₁₄ClN₅ MW: 227.7 g/mol |
MDL Number:
MFCD13561893 CAS Number: 1220016-74-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...