4-Chloro-6-(1-pyrrolidinyl)-1,3,5-triazin-2-ylamine

Supplier: Matrix Scientific

052074-500MG 052074-2.5G
101898-186EA 461.54 USD
101898-186 101898-188
4-Chloro-6-(1-pyrrolidinyl)-1,3,5-triazin-2-ylamine
4-Chloro-6-(1-pyrrolidinyl)-1,3,5-triazin-2-ylamine

Hazard: Irritant

Formula: C₇H₁₀ClN₅
MW: 199.64 g/mol
MDL Number: MFCD09035496
CAS Number: 937670-30-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR