4-(3-Amino-2-pyridinyl)-2-piperazinone
Supplier: Matrix Scientific
052006-500MG
052006-2.5G
101897-914EA
364.62
USD
101897-914
101897-916
4-(3-Amino-2-pyridinyl)-2-piperazinone
4-(3-Amino-2-pyridinyl)-2-piperazinone
Hazard: Irritant
Formula:
C₉H₁₂N₄O MW: 192.22 g/mol |
MDL Number:
MFCD09040654 CAS Number: 926197-03-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...