3-(tert-Butyl)phenyl 4-piperidinylmethyl ether hydrochloride

Supplier: Matrix Scientific

050712-500MG 050712-2.5G
101892-748EA 364.62 USD
101892-748 101892-750
3-(tert-Butyl)phenyl 4-piperidinylmethyl ether hydrochloride
3-(tert-Butyl)phenyl 4-piperidinylmethyl ether hydrochloride

Hazard: Irritant

Formula: C₁₆H₂₆ClNO
MW: 283.84 g/mol
MDL Number: MFCD13561103
CAS Number: 1220033-77-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR