(±)-Propranolol hydrochloride ≥95%

Supplier: Matrix Scientific

Synonyms: 1-(Isopropylamino)-3-(1-naphthyloxy)-2-propanol hydrochloride, Propranolol hydrochloride, DL-Propranolol hydrochloride, (±)-1-Isopropylamino-3-(1-naphthyloxy)-2-propanol hydrochloride

047896-25G 047896-100G
101881-898EA 84.62 USD
101881-898 101881-900
(±)-Propranolol hydrochloride ≥95%
(±)-Propranolol hydrochloride

Purity: 95% CAS: 318-98-9 MDL Number: MFCD00012558 Molecular Formula: C16H22ClNO2 Molecular Weight: 295,81 Hazard: Irritant

Formula: C₁₆H₂₁NO₂·HCl
MW: 295.81 g/mol
MDL Number: MFCD00012558
CAS Number: 318-98-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR