1-(3-Ethoxyphenyl)methanamine
Supplier: Matrix Scientific
037111-500MG
037111-2.5G
101842-700EA
47.69
USD
101842-700
101842-702
1-(3-Ethoxyphenyl)methanamine
1-(3-Ethoxyphenyl)methanamine
Hazard: Irritant
Formula:
C₉H₁₃NO |
MDL Number:
MFCD03410991 CAS Number: 93071-76-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...