5-Methyl-[1,2,4]triazole-3,4-diamine
Supplier: Matrix Scientific
Synonyms:
5-Methyl-4H-1,2,4-triazole-3,4-diamine
020792-500MG
101810-780EA
280
USD
101810-780
5-Methyl-[1,2,4]triazole-3,4-diamine
5-Methyl-[1,2,4]triazole-3,4-diamine
Hazard: Irritant
Formula:
C₃H₇N₅ MW: 113.12 g/mol |
MDL Number:
MFCD03789559 CAS Number: 21532-07-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...