1,1-Dimethylpropargylamine

Supplier: Matrix Scientific

Synonyms: 2-Methylbut-3-yn-2-amine

008052-1G 008052-5G
101793-920EA 193.85 USD
101793-920 101793-922
1,1-Dimethylpropargylamine
1,1-Dimethylpropargylamine

Hazard: Flammable/Corrosive/Stored under Argon/Air Sensitive/Lachrymator

Formula: C₅H₉N
MW: 83.134 g/mol
Boiling Pt: 79…80 °C
Melting Pt: 18 °C
Density: 0.79
Flash Pt: 2 °C
MDL Number: MFCD00008052
CAS Number: 2978-58-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR