1-(4-Fluoro-2-nitrophenyl)piperazine
Supplier: Matrix Scientific
005905-250MG
005905-1G
101789-724EA
993.85
USD
101789-724
101789-726
1-(4-Fluoro-2-nitrophenyl)piperazine
1-(4-Fluoro-2-nitrophenyl)piperazine
Hazard: Irritant
Formula:
C₁₀H₁₂FN₃O₂ |
MDL Number:
MFCD00798371 CAS Number: 243128-46-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...