Methyl perfluorononanoate ≥98%

Supplier: Matrix Scientific

005785-25G 005785-100G
101789-522EA 353.85 USD
101789-522 101789-524
Methyl perfluorononanoate ≥98%
Methyl perfluorononanoate

Hazard: Irritant

Formula: C₁₀H₃F₁₇O₂
MW: 478.11 g/mol
Boiling Pt: 63…65 °C (10 Torr)
Flash Pt: >110 °C
MDL Number: MFCD00153213
CAS Number: 51502-45-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR