6-Bromo-3-chloroisoquinoline ≥96%

Supplier: Matrix Scientific

070402-1G 070402-5G
101944-594EA 500 USD
101944-594 101944-596
6-Bromo-3-chloroisoquinoline ≥96%
6-Bromo-3-chloroisoquinoline

Hazard: Irritant

Formula: C₉H₅BrClN
MDL Number: MFCD08703290
CAS Number: 552331-06-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR