5-Amino-3-methylimidazo[1,2-a]pyridine ≥97%

Supplier: Matrix Scientific

069783-1G 069783-5G
101941-912EA 1047.69 USD
101941-912 101941-914
5-Amino-3-methylimidazo[1,2-a]pyridine ≥97%
5-Amino-3-methylimidazo[1,2-a]pyridine

Hazard: Irritant

Formula: C₈H₉N₃
MDL Number: MFCD10699209
CAS Number: 74420-50-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR