2-Methylpropanethioamide ≥98%

Supplier: Matrix Scientific

Synonyms: Thioisobutyramide

069704-1G 069704-5G 069704-25G
101941-532EA 181.54 USD
101941-532 101941-534 101941-536
2-Methylpropanethioamide ≥98%
2-Methylpropanethioamide

Hazard: Irritant

Formula: C₄H₉NS
Melting Pt: 45…48 °C
MDL Number: MFCD07369538
CAS Number: 13515-65-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR