3-(Tributylstannyl)-4-(trifluoromethyl)-1H-pyrazole ≥90%

Supplier: Matrix Scientific

069688-500MG 069688-1G
101941-450EA 1221.54 USD
101941-450 101941-452
3-(Tributylstannyl)-4-(trifluoromethyl)-1H-pyrazole ≥90%
3-(Tributylstannyl)-4-(trifluoromethyl)-1H-pyrazole

Hazard: Irritant

Formula: C₁₆H₂₉F₃N₂Sn
MDL Number: MFCD10699133
CAS Number: 790661-62-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR