6-(4-Chlorophenyl)pyridazin-3(2H)-one

Supplier: Matrix Scientific

065274-500MG
101929-350EA 364.62 USD
101929-350
6-(4-Chlorophenyl)pyridazin-3(2H)-one
6-(4-Chlorophenyl)pyridazin-3(2H)-one

Hazard: Irritant

Formula: C₁₀H₇ClN₂O
MDL Number: MFCD02676708
CAS Number: 2166-13-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR