3-Chloroquinoxalin-2-amine ≥95%

Supplier: Matrix Scientific

059850-250MG 059850-1G
101919-288EA 153.85 USD
101919-288 101919-290
3-Chloroquinoxalin-2-amine ≥95%
3-Chloroquinoxalin-2-amine

Hazard: Irritant

Formula: C₈H₆ClN₃
MDL Number: MFCD00234398
CAS Number: 34117-90-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR