Tamoxifen citrate

Supplier: Matrix Scientific

Synonyms: (Z)-1-(4-Dimethylaminoethoxyphenyl)-1,2-diphenyl-1-butene Citrate

054648-500MG
101907-994EA 190.77 USD
101907-994
Tamoxifen citrate
Tamoxifen citrate

CAS: 54965-24-1
MDL Number: MFCD00058321
Molecular Formula: C32H37NO8
Molecular Weight: 563,65
Hazard: Irritant

Formula: C₂₆H₂₉NO
MW: 563.65 g/mol
MDL Number: MFCD00058321
CAS Number: 54965-24-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR